Order Entry
Switzerland
ContactUsLinkComponent
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥94% (by HPLC), Calbiochem®, Millipore®
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥94% (by HPLC), Calbiochem®, Millipore®
Catalog # 573115-50
CAS Number:  
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥94% (by HPLC), Calbiochem®, Millipore®
Catalog # 573115-50
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :436.37 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Freezer
CAS Number:92921-24-9
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978