Order Entry
Switzerland
ContactUsLinkComponent
 
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate), Pierce™
Catalog # PIER21857
CAS Number:  
undefined
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate), Pierce™
Catalog # PIER21857
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :312.36 g/mol
Storage Temperature:Freezer
CAS Number:68181-17-9
Formula:C₁₂H₁₂N₂O₄S₂
MDL Number:MFCD00009636