Order Entry
Switzerland
ContactUsLinkComponent
Rat Neuregulin-1/NRG1 ELISA kit (colorimetric)
Rat Neuregulin-1/NRG1 ELISA kit (colorimetric)
Catalog # NOVUNBP2-68071
Supplier:  Novus Biologicals
CAS Number:  
Rat Neuregulin-1/NRG1 ELISA kit (colorimetric)
Catalog # NOVUNBP2-68071
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Description:
    Rat Neuregulin-1/NRG1 ELISA kit (colorimetric)
  • Sample volume:
    100 µl

Specifications

About this item

This kit recognizes Rat NRG-1 in samples. No significant cross-reactivity or interference between Rat NRG-1 and analogues was observed.

  • Standard Curve Range: 0,63 to 40 ng/ml
  • Sensitivity: 0,38 ng/ml
  • Inter-Assay: CV% <4,97%
  • Intra-Assay: CV% <4,89%
  • Spike Recovery: 84 to 106%
  • Kit Type: ELISA Kit (Colorimetric)
  • Gene: NRG1