Order Entry
Switzerland
ContactUsLinkComponent
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester), Pierce™
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester), Pierce™
Catalog # PIER22298
CAS Number:  
undefined
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester), Pierce™
Catalog # PIER22298
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :266.21 g/mol
Storage Temperature:Refrigerator
CAS Number:55750-62-4
Formula:C₁₁H₁₀N₂O₆
MDL Number:MFCD00043141