Order Entry
Switzerland
ContactUsLinkComponent
 
MBS (N-Succinimidyl 3-maleimidobenzoate), Pierce™
Catalog # PIER22311
CAS Number:  
undefined
MBS (N-Succinimidyl 3-maleimidobenzoate), Pierce™
Catalog # PIER22311
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Maleimide, NHS Ester
  • Spacer:
    Short
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

Specification Test Results

About this item

 :
 :314.25 g/mol
CAS Number:58626-38-3
Formula:C₁₅H₁₀N₂O₆
MDL Number:MFCD00005514