Order Entry
Switzerland
ContactUsLinkComponent
BSA-Palmitate Saturated Fatty Acid Complex
BSA-Palmitate Saturated Fatty Acid Complex
Catalog # CAYM29558-5
Supplier:  Cayman Chemical
CAS Number:  
BSA-Palmitate Saturated Fatty Acid Complex
Catalog # CAYM29558-5
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Concentration:
    5 mM
  • Pk:
    5 mL
  • Description:
    BSA-Palmitate Saturated Fatty Acid Complex

Specifications

About this item

BSA-Palmitate saturated fatty acid complex is composed of palmitic acid and bovine serum albumin (BSA) at an approximately 6:1 molar ratio of palmitate:BSA. It can be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring fatty acid oxidation or similar processes in various cellular metabolic studies.

  • A fatty acid substrate
  • Origin: Animal/Bovine
  • Stability: ≥ 1 year

The BSA/BSA-FAs are acceptable for use with short-term cell culture applications (acute treatment to 18 hours), however for long-term applications (25+ hours) the product can/should be filter sterilized using a 0.2 µm filter and sterile receptacle without affecting the performance. For best results, it is recommended that this product be used in conjunction with BSA control for fatty acid complexes (5 mM).

Formulation: 5 mM Palmitate:0.8 mM BSA (6:1 palmitate:BSA) in 150 mM sodium chloride, pH 7.4

Caution: Store at –20 °C.