Order Entry
Switzerland
ContactUsLinkComponent
Human IL18BPa protein (His Tag)
Human IL18BPa protein (His Tag)
Katalognr. SINO10357H08H-200
Supplier:  Sino Biological
CAS Number:  
Human IL18BPa protein (His Tag)
Katalognr. SINO10357H08H-200
Supplier:  Sino Biological
CAS Number:  

Specifications

  • Pk:
    200 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20…–80 °C
  • Endotoxin content:
    <1,0 EU per μg of the protein as determined by the LAL method
  • Biological activity:
    1. Measured by its ability to bind biotinylated Cynomolgus IL18 in functional ELISA.
    2. Measured by its ability to inhibit the IL-18-induced response of KG‑1 human acute myelogenous leukemia cells. The ED50 for this effect is 0,05 - 0,35 μg/ml in the presence of 50 ng/ml of recombinant human IL-18.
  • Gene ID:
    NP_001034748.1
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    IL18BPa
  • Protein/peptide name:
    IL18BP
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant human IL18BPa consists of 175 amino acids and has a predicted molecular mass of 19 kDa.In SDS-PAGE under reducing conditions, the apparent molecular mass of human IL18BPa is approximately 40,3 kDa due to glycosylation.
  • Sequence:
    Met1-Gly194
  • Formulation:
    Powder
  • Shipping temperature:
    Ambient

Specifications

About this item

A DNA sequence encoding the human IL18BPa (NP_001034748.1) (Met1-Gly194) was expressed with the polyhistidine tag at the C-terminus.