ContactUsLinkComponent
Elbow fitting system waste cap, b.safe
Elbow fitting system waste cap, b.safe
Catalog # BOHLR1227-01
Supplier:  Bohlender
undefined
Elbow fitting system waste cap, b.safe
Catalog # BOHLR1227-01
Supplier:  Bohlender
Specifications
  • Thread:
    GL 25 (male), GL 25 (female)
  • Description:
    Elbow fitting system waste cap
  • Pk:
    1
Specifications
About this item
Screw cap for thread GL 25 made of PPS with rotary elbow fitting with male thread GL 25 and drip edge at female thread GL 25.
  • Made of static dissipative PE-EX
  • Space-saving
  • Without sagging
An angle of approx. 93° allows a connection of b.safe Waste Tubes to b.safe system waste caps. The position of the tube connection is freely selectable due to the rotary elbow fitting. The drip edge in the screw cap GL 25 ensures that even small amounts of solvents can be lead into the waste canister without leakage.