Order Entry
Switzerland
ContactUsLinkComponent
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 216254-1
CAS Number:  
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 216254-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :808.69 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969