Order Entry
Switzerland
ContactUsLinkComponent
Canine EGFR / HER1 / ErbB1 protein (His Tag)
Canine EGFR / HER1 / ErbB1 protein (His Tag)
Catalog # SINO70026-D08B-50
Supplier:  Sino Biological
CAS Number:  
undefined
Canine EGFR / HER1 / ErbB1 protein (His Tag)
Catalog # SINO70026-D08B-50
Supplier:  Sino Biological
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Dog
  • Storage conditions:
    –20…–80 °C
  • Endotoxin content:
    <1,0 EU per μg of the protein as determined by the LAL method
  • Biological activity:
    Measured by its binding ability in a functional ELISA.
    Immobilized canine EGFR-His at 10 μg/ml (100 μl/well) can bind biotinylated canine His-EGF(Cat:70013-D07E), The EC50 of biotinylated canine His-EGF(Cat:70013-D07E) is 0,16 - 0,36 μg/ml
  • Gene ID:
    XP_533073.3
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    EGFR
  • Protein/peptide name:
    EGFR
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant canine EGFR comprises 629 amino acids and has a predicted molecular mass of 69,8 kDa. The apparent molecular mass of the protein is approximately 87,2 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Ser618
  • Formulation:
    Powder
  • Shipping temperature:
    Ambient

Specifications

About this item

A DNA sequence encoding the canine EGFR (XP_533073.3) (Met1-Ser618) was expressed with a C-terminal polyhistidine tag.