ContactUsLinkComponent
Vaporised hydrogen peroxide tri-metric chemical indicator, Attest™
Vaporised hydrogen peroxide tri-metric chemical indicator, Attest™
Catalog # MMME1348
Supplier:  3M MEDICAL
Vaporised hydrogen peroxide tri-metric chemical indicator, Attest™
Catalog # MMME1348
Supplier:  3M MEDICAL
Specifications
  • Description:
    Attest™ Vaporized hydrogen peroxide tri-metric chemical indicator
  • Pk:
    1.000
Specifications
About this item
The 3M™ Attest™ Vaporized Hydrogen Peroxide Tri-Metric Chemical Indicator 1348/1348E verifies that the stated values for the three critical parameters of exposure time, temperature, and amount of vaporized hydrogen peroxide have been achieved within a package or containment device (i.e. wrapped trays, rigid containers, sterilization pouches, and other types of packs) and/or at a specific location within the load or empty chamber.
  • Detects failed VH2O2 sterilization cycles
  • Won’t change color before sterilization under ambient light
  • Consistent blue-toward-pink color change for vaporized hydrogen peroxide
  • Proprietary protective film prevents ink smears on surgical instruments
The 3M™ Attest™ Vaporized Hydrogen Peroxide Tri-Metric Chemical Indicator 1348/1348E is a Type 4 indicator as categorized by ISO 11140-1:2014.  The 3M™ Attest™ Tri-Metric 1348/1348E CI is the first and only CI in the FDA’s newly-created multivariable indicator category for chemical vapor sterilisation, as it independently verifies that the stated values of the three critical parameters of exposure time, temperature, and concentration of vaporized hydrogen peroxide have been met.
Upon exposure to vaporized hydrogen peroxide, the color of the coated indicator strip progressively changes from blue toward pink along the strip. The progression of the blue to pink color change along the strip is visible through a window with marked 'REJECT' and 'ACCEPT' zones. The extent of the progression depends on exposure time, temperature, and amount of vaporized hydrogen peroxide.