Mouse DLL1 / Delta-1 Protein (His Tag)
Catalog # SINO50522-M08H-50
Supplier: Sino Biological
CAS Number:
Specifications
- Pk:50 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage conditions:–20…–80 °C
- Endotoxin content:< 1.0 EU per μg of the protein as determined by the LAL method
- Biological activity:1. Measured by its ability to bind human NOTCH1 in a functional ELISA. 2. Measured by the ability of the immobilized protein to enhance BMP2-induced alkaline phosphatase activity in C3H10T1/2 mouse embryonic fibroblast cells. The ED50 for this effect is typically 2-20 μg/mL.
- Gene ID:NP_031891.2
- Reconstitution instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Animal-free:Y
- Protein/peptide name:DLL1
- Purity:> 95 % as determined by SDS-PAGE
- Molecular weight:The recombinant mouse DLL1 consists of 506 amino acids and has a predicted molecular mass of 55 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of rm DLL1 is approximately 65 kDa due to glycosylation.
- Sequence:Met1-Gln516
- Formulation:Lyophilized from sterile PBS, pH 7.4, 20% glycerol
- Shipping temperature:In general, recombinant proteins are provided as lyophilized powder which are shipped at ambient temperature. Bulk packages of recombinant proteins are provided as frozen liquid. They are shipped out with blue ice unless customers require otherwise.
Specifications
About this item
A DNA sequence encoding the extracellular domain of mouse DLL1 (NP_031891.2) (Met 1-Gln 516) was expressed, with a polyhistidine tag at the C-terminus.