Order Entry
Switzerland
ContactUsLinkComponent
Mouse TNFR1 / CD120a / TNFRSF1A Protein (Fc Tag)
Mouse TNFR1 / CD120a / TNFRSF1A Protein (Fc Tag)
  SINO50496-M02H-100
 :  
undefined
Mouse TNFR1 / CD120a / TNFRSF1A Protein (Fc Tag)
  SINO50496-M02H-100
 :  

 

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20…–80 °C
  • Endotoxin content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological activity:
    1. Measured by its binding ability in a functional ELISA. Immobilized Mouse TNF-alpha (Cat:50349-MNAE) at 2 μg/ml (100 μl/well) can bind Mouse TNFRSF1AhFc (Cat:50496-M02H), the EC50 of Mouse TNFRSF1AhFc is 1.5-9.0 ng/ml.
    2. Measured by its ability to inhibit the TNFα-mediated cytotoxicity in the L-929 mouse fibroblast cells in the presence of the metabolic inhibitor actinomycin D. The ED50 for this effect is typically 15-65 ng/ml in the presence of 0.1 ng/ml of recombinant mouse TNFα.
  • Gene ID:
    NP_035739.2
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD120a,FPF,p55,p55-R,TNF-alphaR1,TNF-R,TNF-R-I,TNF-R1,TNF-R55,TNFalpha-R1,TNFAR,Tnfr-2,Tnfr1,TNFR60,TNFRI,TNFRp55
  • Protein/peptide name:
    TNFRSF1A
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    The secreted recombinant mouse TNFRSF1A/Fc is a disulfide-linked homodimer. The reduced monomer consists of 424 amino acids and has a predicted molecular mass of 47.4 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of rm TNFRSF1A/Fc monomer is approximately 55 kDa due to glycosylation.
  • Sequence:
    Met1-Ala212
  • Formulation:
    Powder
  • Shipping temperature:
    Ambient

 

 

A DNA sequence encoding the mouse TNFRSF1A (NP_ 035739.2) extracellular domain (Met 1-Ala 212) was fused with the Fc region of human IgG1 at the C-terminus.