Human IL1RL2 / IL-1Rrp2 Protein (His Tag)
Katalognr. SINO14398-H08H-100
Anbieter: Sino Biological
CAS-Nummer:
Technische Daten
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20…–80 °C
- Endotoxin content:<1,0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_003845.2
- Reconstitution instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Animal-free:Y
- Protein synonyms:IL-1Rrp2,IL-36R,IL1R-rp2,IL1RRP2
- Protein/peptide name:IL1RL2
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:The recombinant human IL1RL2 consists 329 amino acids and predicts a molecular mass of 37,6 kDa.
- Sequence:Met1-Tyr337
- Formulation:Powder
- Shipping temperature:Ambient
Technische Daten
Über diesen Artikel
A DNA sequence encoding the human PTPN12 (AAA36529.1) (Met1-Gln355) was expressed and purified with two additional amino acids (Gly & Pro) at the N-terminus.