Order Entry
Switzerland
ContactUsLinkComponent
 
Human STAT6 Protein (His Tag)
  SINO13190-H08B-200
 :  
undefined
Human STAT6 Protein (His Tag)
  SINO13190-H08B-200
 :  

 

  • Pk:
    200 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20…–80 °C
  • Endotoxin content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological activity:
    0
  • Gene ID:
    P42226-1
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    D12S1644,IL-4-STAT,STAT6B,STAT6C
  • Protein/peptide name:
    STAT6
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant human STAT6 consists of 857 amino acids and predicts a molecular mass of 95.5 kDa. It migrates as an approximately 100 kDa band in SDS-PAGE in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Trp847
  • Formulation:
    Powder
  • Shipping temperature:
    Ambient

 

 

A DNA sequence encoding the extracellular domain (Met-Pro 250) of human erythropoietin receptor (NP_000112.1) precursor was expressed with the C-terminal fused Fc region of human IgG1.