Anti-Her2 / ERBB2 Rabbit Polyclonal Antibody
SINO50714-T48-200
- Antibody type:Primary
- Antigen symbol:Her2 / ERBB2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Storage temperature:Can be stored at 2℃-8℃. Stable for twelve months at -20℃ to -80℃. Sodium azide is recommended to avoid contamination (final concentration 0.05%-0.1%). It is toxic to cells and should be disposed of properly. Avoid repeated freeze-thaw cycles.
- Shipping temperature:ambient temperature
- Size:200 µl
- Pk:200 µl
Anti-Her2 / ERBB2 antibody
Type: Primary
Antigen: Her2/ERBB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse