Multiple affinity removal spin cartridge - Ms-3 (mouse)
Multiple affinity removal spin cartridge - Ms-3 (mouse), 0,45 ml
Catalog # AGLS5188-5289
Supplier: AGILENT
Jump to:
Product Details & Documents
The multiple affinity removal spin cartridge – Mouse 3 is designed as a simple way to remove three interfering high-abundant proteins from mouse plasma samples. Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
- Cartridge eliminates approximately 80% of total protein (albumin, IgG, and transferrin)
- Serum load volume ranges between 25 to 30 µl per run
- Total load volume is 200 µl (serum plus buffer)
- Minimize sample loss by utilizing our rapid, single injection protocols and optimized buffers
- Cartridges can last for over 200 injections
- Process the most plasma/serum over the column lifetime, up to 7800 µl
Protocols and column are optimised for use with mouse serum. Column not recommended for use with human serum. Compatible with standard benchtop microcentrifuges and luer-lok syringes.
Specifications
- Capacity:0,45 ml
- Description:Multiple affinity removal spin cartridge - Ms-3 (mouse)



