InterPlay adenoviral TAP system
Catalog # AGLS240101
Anbieter: AGILENT
CAS-Nummer:
Technische Daten
- VE:60 µG
- Bezeichnung:InterPlay N-Terminal Mammalian TAP vectors, 3×20 μg
Technische Daten
Über diesen Artikel
The InterPlay Mammalian TAP system allows you to recover interacting proteins from mammalian cells.
- Protease digestion unnecessary to recover intact interacting protein partners since SBP and CBP elute easily from their affinity resins
- Exceptionally pure protein for downstream analysis
This method is based on expression of a protein of interest fused to two affinity tags: a streptavidin binding peptide (SBP) and a calmodulin binding peptide (CBP). Tandem affinity purification yields your tagged protein and interacting proteins using gentle washing and small molecule elution conditions.
Caution: For research use only. Not for use in diagnostic procedures.