Multiple affinity removal column human 14
Supplier: AGILENT
Jump to:
About this item
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
- Made of PEEK (polyetheretherketone)
- High-capacity serum load
- Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
2 Options Available Below
Product Details & Documents
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
- Made of PEEK (polyetheretherketone)
- High-capacity serum load
- Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Product Family Options
Product Information
- Ø int.LengthAvailabilityPrice
- AGLS5188-6557Multiple affinity removal column, human-14Multiple affinity removal column, human-144,60 mm50 mmLog in for availability1 * 1 STSFr. 7’025.00
- AGLS5188-6558Multiple affinity removal column, human-14Multiple affinity removal column, human-144,60 mm100 mmLog in for availability1 * 1 STSFr. 11’241.00