Anti-TNFA mouse monoclonal antibody [clone: 9A7A5G9]
SINO10602-MM01-100
- Antibody type:Primary
- Antigen symbol:TNFA
- Clonality:Monoclonal
- Clone:Clone: 9A7A5G9
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Seq:02
- Form:liquid
- Storage temperature:Can be stored at 2...8 ℃. Stable for twelve months at −20...−80 ℃. Sodium azide is recommended to avoid contamination (final concentration 0,05% - 0,1%). It is toxic to cells and should be disposed of properly. Avoid repeated freeze-thaw cycles.
- Shipping temperature:ambient temperature
- Pk:100 µl
Anti-TNF-alpha antibody
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: Clone: 9A7A5G9
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human