Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-DUSP10 rabbit polyclonal antibody
  NOVUNBP1-31001
Anti-DUSP10 rabbit polyclonal antibody
  NOVUNBP1-31001

 

  • Antibody type:
    Primary
  • Antigen name:
    Dual Specificity Protein Phosphatase 10
  • Antigen symbol:
    DUSP10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    11221
  • Antigen synonyms:
    MKP5serine/threonine specific protein phosphatase|dual specificity phosphatase MKP-5|dual specificity phosphatase 10|MAP kinase phosphatase 5|MKP-5dual specificity protein phosphatase 10|EC 3.1.3.16|EC 3.1.3.48|Mitogen-activated protein kinase phosphatase 5
  • Storage buffer:
    PBS (pH 7,0) and 40% Glycerol
  • Storage temperature:
    Aliquot and store at −20 or −80 °C. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human DUSP10. The exact sequence is proprietary.
  • Purification:
    Immunogen affinity purified
  • Pk:
    100 µl

 

 

The DUSP10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DUSP10. This antibody reacts with human. The DUSP10 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: DUSP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human