Anti-collagen I alpha 1 rabbit polyclonal antibody
Katalognr. NOVUNBP1-30054
Anbieter: Novus Biologicals
Technische Daten
- Antibody type:Primary
- Antigen name:Collagen I alpha 1
- Antigen symbol:COL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Sheep,Human,Rat,Mouse
- Western blot:Yes
- Gene ID:1277
- Antigen synonyms:Alpha-1 type I collagen|alpha 1|Collagen 1|OI4|Collagen1|collagen of skin|collagen alpha-1(I) chain|collagen|pro-alpha-1 collagen type 1|Collagen 1 alpha 1|Collagen-1|alpha-1 chain|collagen alpha 1 chain type I|tendon and bone|type I
- Storage buffer:PBS
- Storage temperature:Store at −20 °C. Avoid freeze-thaw cycles.
- Concentration:1,0 mg/ml
- Immunogen:Synthetic peptide from the C-terminal region of human collagen I alpha1 telopeptide sequence.
- Purification:Immunogen affinity purified
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
The Collagen I alpha 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Collagen I alpha 1. This antibody reacts with human, mouse, rat, sheep. The Collagen I alpha 1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: Collagen I alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Sheep