Anti-DEC1 rabbit polyclonal antibody
100 μl
Catalog # NOVUNB100-1800
Supplier: Novus Biologicals
Jump to:
Product Details & Documents
The DEC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to 43070. This antibody reacts with human, mouse. The DEC1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Simple Western
Specifications
- Size:100 μl
- Western blot:Yes
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:Immunogen affinity purified
- Antibody type:Primary
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Host:Rabbit
- Concentration:1,0 mg/ml
- Cat. No.:NOVUNB100-1800
- Reactivity:Human|Mouse
- Gene ID:8553
- Antigen synonyms:SHARP-2|Class B basic helix-loop-helix protein 2|bHLHb2|Stra14|member e40|basic helix-loop-helix domain containing|bHLHe40|differentiated embryo chondrocyte expressed gene 1|class B|Stimulated by retinoic acid gene 13 protein|Differentially expressed in chondrocytes protein 1|differentially expressed in chondrocytes 1|SHARP2|basic helix-loop-helix family|STRA13FLJ99214|DEC1HLHB2|2|Enhancer-of-split and hairy-related protein 2|class E basic helix-loop-helix protein 40
- Immunogen:Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503]
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:DEC1
- Storage buffer:PBS



