Human recombinant STING R224 variant (from E. coli)
CAYM23593-100
:
- Pk:100 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Storage conditions:–80 °C
- Shipping conditions:–80 °C, dry ice
- Enzyme name:STING R224 variant
- Enzyme synonyms:Endoplasmic Reticulum Interferon Stimulator, Stimulator of Interferon Genes, TMEM173
- Purity:≥70% estimated by SDS-PAGE
- Molecular weight:28,8 kDa
- Formulation:50 mM HEPES, pH 8,0, 150 mM sodium chloride, and 10% glycerol