Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant TBK1 (from Baculovirus (Insect cells))
Human recombinant TBK1 (from Baculovirus (Insect cells))
Catalog # CAYM22817-50
Supplier:  Cayman Chemical
CAS Number:  
undefined
Human recombinant TBK1 (from Baculovirus (Insect cells))
Catalog # CAYM22817-50
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage conditions:
    –80 °C
  • Shipping conditions:
    –80 °C, dry ice
  • Enzyme name:
    TBK1
  • Purity:
    ≥50% as estimated by SDS-PAGE
  • Molecular weight:
    84 kDa
  • Formulation:
    50 mM HEPES, pH 8,0, 150 mM sodium chloride, and 10% glycerol

Specifications

About this item