Order Entry
Switzerland
ContactUsLinkComponent
Lysing matrix Y, FastPrep™, MP Biomedicals
Lysing matrix Y, FastPrep™, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Lysing matrix Y, FastPrep™, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Catalog No:
  • 116960001.
  • 116960010.
  • 116975025.
  • 116975050.
  • Specific gravity:
  • 6,02
  • 6,02
  • 6,02
  • 6,02
  • Size range:
  • 0,4 - 0,6 mm
  • 0,4 - 0,6 mm
  • 0,4 - 0,6 mm
  • 0,4 - 0,6 mm
  • Storage and handling:
  • Store at Room Temperature (15 - 30 °C)
  • Store at Room Temperature (15 - 30 °C)
  • Store at Room Temperature (15 - 30 °C)
  • Store at Room Temperature (15 - 30 °C)
  • Sample type:
  • Bacteria (Gram +/-), Yeast, Fungi, Spores, Algae, Biofilms
  • Bacteria (Gram +/-), Yeast, Fungi, Spores, Algae, Biofilms
  • Bacteria (Gram +/-), Yeast, Fungi, Spores, Algae, Biofilms
  • Bacteria (Gram +/-), Yeast, Fungi, Spores, Algae, Biofilms
  • Molecule enriched:
  • DNA, RNA, Proteins, Other small molecules
  • DNA, RNA, Proteins, Other small molecules
  • DNA, RNA, Proteins, Other small molecules
  • DNA, RNA, Proteins, Other small molecules
  • Materials required (not supplied):
  • FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
  • FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
  • FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
  • FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
  • Material of construction:
  • 0,5 mm diameter Yttria-Stabilized Zirconium Oxide beads
  • 0,5 mm diameter Yttria-Stabilized Zirconium Oxide beads
  • 0,5 mm diameter Yttria-Stabilized Zirconium Oxide beads
  • 0,5 mm diameter Yttria-Stabilized Zirconium Oxide beads
  • Instrument Compatibility:
  • FastPrep-96
  • FastPrep-96
  • FastPrep-24, FastPrep-96, FastPrep-24 5G
  • FastPrep-24, FastPrep-96, FastPrep-24 5G
  • Hardness:
  • Mohs 9, Rockwell A >87, Vickers >1350
  • Mohs 9, Rockwell A >87, Vickers >1350
  • Mohs 9, Rockwell A >87, Vickers >1350
  • Mohs 9, Rockwell A >87, Vickers >1350
  • Adapter compatibility:
  • 96-Well Adapter
  • 96-Well Adapter
  • TeenPrep (12×15 ml), CoolTeenPrep (6×15 ml), TeenFlex (24×15 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
  • TeenPrep (12×15 ml), CoolTeenPrep (6×15 ml), TeenFlex (24×15 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
  • Application areas:
  • Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
  • Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
  • Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
  • Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation

Specifications

Frequently Bought Together