Human Native Collagen IV alpha 1
Katalognr. NOVUNBP1-97268
Supplier: Novus Biologicals
CAS Number:
Specifications
- Pk:0,5 mg
- Conjugation:Unconjugated
- Protein/peptide type:Native
- Species:Human
- Storage conditions:Store at 4 °C. Do not freeze.
- Gene ID:1282
- Protein synonyms:alpha 1|COL4A1 NC1 domain|collagen alpha-1(IV) chain|collagen|arresten|EC 3.4.23|Collagen-4|alpha-1 chain|type IV|collagen IV|alpha-1 polypeptide|EC 6.3.1.2
- Protein/peptide name:Collagen IV alpha 1
- Purity:SDS-PAGE
- Concentration:1,0 mg/ml
Specifications
About this item
The Collagen IV alpha 1 Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.