Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Multiple affinity removal columns - Mouse 3

Multiple affinity removal columns - Mouse 3

4,60 mm, 50 mm
Catalog # AGLS5188-5217
Supplier:   AGILENT
Multiple affinity removal columns - Mouse 3
Catalog # AGLS5188-5217
Supplier:  AGILENT
Multiple affinity removal columns - Mouse 3
Multiple affinity removal columns - Mouse 3

4,60 mm, 50 mm

Price
SFr. 4’721.00
Log in for availability
1 * 1 ST
Product Details & Documents

The multiple affinity removal LC column – Mouse 3 is designed as a simple way to remove three interfering high-abundant proteins from mouse plasma samples.

Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.

Caution: For research use only.

Specifications
  • Ø int.:
    4,60 mm
  • Length:
    50 mm
  • Seq:
    01