Order Entry
Switzerland
ContactUsLinkComponent
Phenolphthalein 0.375% in methanol indicator
Flammable
Toxic
Health
Phenolphthalein 0.375% in methanol indicator
Catalog # 86196.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Toxic
Health
Phenolphthalein 0.375% in methanol indicator
Catalog # 86196.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Application test:
    Passes test

Specification Test Results

About this item

 :
 :318.33 g/mol
Storage Temperature:Ambient
UN:1230
ADR:3 (6.1),II
 :GHS02
 :GHS06
 :GHS08
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913