ContactUsLinkComponent
Supplied air system, V-series, Versaflo™
Supplied air system, V-series, Versaflo™
Codice catalogo MMMAS212
Nome del fornitore:  3M
undefined
Supplied air system, V-series, Versaflo™
Codice catalogo MMMAS212
Nome del fornitore:  3M
Specifiche
  • Description:
    Supplied air system, V-series
  • Seq:
    02
  • Pk:
    1
Specifiche
Informazioni su questo articolo
Regulator Assembly S-212 is a replacement part for use with Versaflo™ Supplied Air System S-200+.
  • Spare or replacement part
  • Designed for use with 3M™ Versaflo™ Supplied Air System S-200+
  • CE approved
Prolong the service life of the 3M™ Versaflo™ Supplied Air System S-200+ with 3M™ Regulator Assembly, S-212. This spare regulator is specially designed for S-200+ systems.