ContactUsLinkComponent
Powered air respirator head, neck, and shoulder covers, Versaflo™
Powered air respirator head, neck, and shoulder covers, Versaflo™
Catalog # MMMAM972
Supplier:  3M
undefined
Powered air respirator head, neck, and shoulder covers, Versaflo™
Catalog # MMMAM972
Supplier:  3M
Specifications
  • Colour:
    Black
  • Description:
    Head, neck, and shoulder covers
  • Pk:
    1
Specifications
About this item
The Versaflo™ Headtop Cover M-972 is a flame-resistant polyester headtop cover designed to protect the M-Series headtop from sparks and hot particles.
  • Flame-resistant headtop cover
  • Use with any Versaflo™ M-Series product
Protect the outside surface of the helmet shell from sparks and hot particles with the Versaflo™ Flame Resistant Headtop Cover, M-972. This cover is made from flame-resistant polyester and is a disposable item. It should be discarded when it becomes damaged or soiled.