ContactUsLinkComponent
Powered and supplied air sweatbands and comfort pads, Versaflo™
Powered and supplied air sweatbands and comfort pads, Versaflo™
Catalog # MMMAM956
Supplier:  3M
undefined
Powered and supplied air sweatbands and comfort pads, Versaflo™
Catalog # MMMAM956
Supplier:  3M
Specifications
  • Description:
    Sweatbands and comfort pads
  • Pk:
    10
Specifications
About this item
The Versaflo™ Size Reducing Ratchet Comfort Pad M-956 is a ratchet-style comfort pad that fits to the back of a 3M™ Versaflo™ M-Series Headtop to reduce the size of the headtop.
  • Size reducing comfort pad fits to rachet of helmet
  • Adds comfort and helps fit smaller wearers
  • Designed for use with 3M™ Versaflo™ M-Series Headtops
Adjust the fit of 3M™ Versaflo™ M-Series Headtop with a Versaflo™ Size Reducing Ratchet Comfort Pad, M-956. It’s easy to fit comfort pads onto the rachet at the back of a helmet to help fit smaller wearers or provide additional comfort to any user.