ContactUsLinkComponent
Powered and supplied air peel off visor covers and films, Versaflo™
  MMMAM928
 :  3M
undefined
Powered and supplied air peel off visor covers and films, Versaflo™
  MMMAM928
 :  3M
  • Colour:
    Transparent
  • Description:
    Peel off visor covers and films
  • Pk:
    40
The Versaflo™ Peel-Off Visor Cover M-928 is an easy-to-use, transparent, self-adhesive visor cover designed to protect 3M™ Versaflo™ M-Series Headtop visors from scratches and overspray of paint or chemicals.
  • Fits over visor, protecting it from scratching or overspray of paint and chemicals
  • Self-adhesive can be easily applied and peeled off
  • Up to five covers can be attached, one on top of the other
The transparent visor cover is self-adhesive and can be applied and peeled off easily. Apply up to five covers at one time, one on top of the other.