Order Entry
Switzerland
ContactUsLinkComponent
AxyMats 24-well sealing mat for deep well plates, non sterile, Axygen®
AxyMats 24-well sealing mat for deep well plates, non sterile, Axygen®
Catalog # CORNAM-24-SQ
Supplier:  Corning
undefined
AxyMats 24-well sealing mat for deep well plates, non sterile, Axygen®
Catalog # CORNAM-24-SQ
Supplier:  Corning
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Colour:
    White
  • Description:
    Autoclavable sealing mat
  • Material:
    Silicone
  • Environmentally Preferable:
  • Pk:
    50

Specifications

About this item

Axygen® AxyMats™ sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.

  • Pierceable Sealing Mat
  • For use with 24 well deep well plates