ContactUsLinkComponent
Masterflex® Union Fittings, Hose Barb, Elbow, PVDF, Avantor®
Masterflex® Union Fittings, Hose Barb, Elbow, PVDF, Avantor®
Certificates
undefined
Masterflex® Union Fittings, Hose Barb, Elbow, PVDF, Avantor®
Certificates
Specifications
  • Model:
  • 30703-28
  • 30703-25
  • Orifice dimensions:
  • 8,73 mm (0,34375")
  • 0,09375"
  • 3,175 mm (0,125")
  • 0,0625"
  • 3,175 mm (0,125")
  • 0,09375"
  • 19,84 mm (0,78125")
  • 14,29 mm (0,5625")
  • 8,73 mm (0,34375")
  • 5,56 mm (0,21875")
  • 11,9 mm (0,46875")
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ETO
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO