ContactUsLinkComponent
Masterflex® L/S® Single-Use High-Performance Sanitary Tubing Assembly, Gamma Irradiated, Avantor®
Masterflex® L/S® Single-Use High-Performance Sanitary Tubing Assembly, Gamma Irradiated, Avantor®
Certificates
undefined
Masterflex® L/S® Single-Use High-Performance Sanitary Tubing Assembly, Gamma Irradiated, Avantor®
Certificates
Specifications
  • Model:
  • 31201-38
  • 31201-39
  • 31201-42
  • Length per pack:
  • 0,46 m (1,5 ft.)
  • 0,91 m (3 ft.)
  • 0,91 m (3 ft.)
  • Max. temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Min. temperature:
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • Pump series:
  • L/S
  • L/S
  • L/S
  • Sterilisation:
  • Autoclave, EtO, Gamma Radiation
  • Autoclave, EtO, Gamma Radiation
  • Autoclave, EtO, Gamma Radiation
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • Yes
  • Yes
  • Yes
  • Warranty:
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect