ContactUsLinkComponent
Masterflex® Color-Coded Sanitary Clamp Fittings, Nylon, Avantor®
Masterflex® Color-Coded Sanitary Clamp Fittings, Nylon, Avantor®
Certificates
undefined
Masterflex® Color-Coded Sanitary Clamp Fittings, Nylon, Avantor®
Certificates
Specifications
  • Model:
  • 31201-45
  • 31201-44
  • 31201-43
  • 31201-48
  • 31201-46
  • Max. operating pressure:
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • 120 psi
  • Sterilisation:
  • Gamma Irradiation, Electron Bean Irradiation, Autoclave, Ethylene Oxide (EtO)
  • Gamma Irradiation, Electron Bean Irradiation, Autoclave, Ethylene Oxide (EtO)
  • Gamma Irradiation, Electron Bean Irradiation, Autoclave, Ethylene Oxide (EtO)
  • Gamma Irradiation, Electron Bean Irradiation, Autoclave, Ethylene Oxide (EtO)
  • Gamma Irradiation, Electron Bean Irradiation, Autoclave, Ethylene Oxide (EtO)
  • Warranty:
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect