ContactUsLinkComponent
Masterflex® Microscope Slides with Printed Sections, Avantor®
Masterflex® Printed Microscope Slide, ten rectangular sections; 144/pk
Catalog # MFLX75955-31
undefined
Masterflex® Printed Microscope Slide, ten rectangular sections; 144/pk
Catalog # MFLX75955-31
Specifications
  • Description:
    Microscope slides, printed with mask, ten rectangular sections
  • L×W:
    76,2×25,4 mm
  • Pk:
    144
  • Slide type:
    Printed without Mask
Specifications
About this item
Optimize sample testing and viewing.
  • Screen-printed section design keeps samples contained
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
Printed microscope slides are available with a variety of section shapes, sizes, counts, colors, and labels. The PTFE-based epoxy ink creates a hydrophobic barrier, holding samples in place for easy viewing under a microscope, and keeping reactions isolated from each other in multi-section slides.
  • Description:
    Microscope slides, printed with mask, ten rectangular sections
  • L×W:
    76,2×25,4 mm
  • Environmentally Preferable:
  • Pk:
    144
  • Slide type:
    Printed without Mask