ContactUsLinkComponent
Liquid Petroleum Gas Corrosion Test Cylinder
L-K Industries LK-TB200 Liquid Petroleum Gas Corrosion Test Cylinder, 1/ea
Catalog # MFLX17700-85
L-K Industries LK-TB200 Liquid Petroleum Gas Corrosion Test Cylinder, 1/ea
Catalog # MFLX17700-85
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Liquid Petroleum Gas Corrosion Test Cylinder, 1/ea
  • Material:
    Stainless steel
  • Compatible with:
    L-K Industries LPG Corrosion Test Cylinder Heater
  • Pk:
    1
Specifications
About this item
Required for use with LPG Corrosion Test Cylinder Heater.
  • Conforms to ASTM D1838 and NPGA Publication 2140 standards
  • Constructed of durable stainless steel for noncorrosive product applications
  • Designed especially for conducting ASTM and NGPA copper strip corrosion tests of liquefied petroleum gas
Compatible with the L-K Industries LP Gas Corrosion Test Cylinder Heater and in compliance with industry standard ASTM D1838. It is made without welds and has an O-ring seal, which will maintain pressure when tightened by hand. No wrenches or other tools are required, so the unit can be used anywhere onsite. The placement of the O-ring, a common replacement part, allows for easy access and reduces down time. It is design tested to 3000 psi.
L-K Industries LP Gas Corrosion Test Cylinder Heater are available as models 17700-40 and -41, and are sold separately.
  • Description:
    Liquid Petroleum Gas Corrosion Test Cylinder, 1/ea
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Compatible with:
    L-K Industries LPG Corrosion Test Cylinder Heater
  • Pk:
    1