Order Entry
Switzerland
ContactUsLinkComponent
Centurion Oil and Petroleum Testing Heated Centrifuge Rotor
Centurion Swing-Out Rotor for Heated Oil Centrifuge
Catalog # MFLX17430-20
Centurion Swing-Out Rotor for Heated Oil Centrifuge
Catalog # MFLX17430-20
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Swing-Out Rotor for Heated Oil Centrifuge
  • Pk:
    1
  • Max. RPM:
    4000 rpm
  • Rotor type:
    Swing-Out
  • Max. Ø of tubes:
    205 mm
  • Warranty:
    3 years

Specifications

About this item

Versatile rotor allows expansion into more Standard Methods.

  • Requires buckets 17430-30, tube adapters, and tube supports (order separately)
  • Description:
    Swing-Out Rotor for Heated Oil Centrifuge
  • Environmentally Preferable:
  • Pk:
    1
  • Max. RPM:
    4000 rpm
  • Rotor type:
    Swing-Out
  • Max. Ø of tubes:
    205 mm
  • Warranty:
    3 years