Order Entry
Switzerland
ContactUsLinkComponent
Masterflex® L/S® Pump Power Supplies, Avantor®
Masterflex® L/S® Power Supply, 90 to 260 VAC Input, 24 VDC Output
Catalog # MFLX07525-90
undefined
Masterflex® L/S® Power Supply, 90 to 260 VAC Input, 24 VDC Output
Catalog # MFLX07525-90

Specifications

  • Description:
    Power Supply, 90 to 260 VAC Input, 24 VDC Output
  • Warranty:
    1 year
  • Pk:
    1

Specifications

About this item

Optimize performance of your Masterflex pump.

  • Suitable for power requirements 90 to 260 VAC

This replacement power supply is suitable for use with the Masterflex C/L Digital pump.

  • Description:
    Power Supply, 90 to 260 VAC Input, 24 VDC Output
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1