About this item
Ideal for installations where there is no gravity drainage available.
- Can be used with other fluids compatible with the wetted materials of construction
- Pump and switch only—for replacing failed pumps in existing tank and shallow pan
- Low-profile system—for lower flows up to 80 GPH
- Slim-footprint systems—come with an oil-resistant tank and check valve
- Large-volume systems—for applications that need larger flow ranges around 200 to 450 GPH
Applications include but are not limited to:
Condensate removal from air conditioners
Drain line collection points
Disposal of wastewater from various water usage devices
Choose from one of the following categories:
Pump and switch only — for replacing failed pumps in existing tank and shallow pan
Low profile systems — lower flows up to 80 GPH
Slim footprint — includes an oil resistant tank and check valve
Large 1 gallon tank — applications that need larger flow ranges around 200 to 450 GPH
Can be used with other fluids compatible with the wetted materials of construction.
- Environmentally Preferable:
Some Products May Appear Restricted