ContactUsLinkComponent
High-Pressure Diaphragm Pumps and Integrated Coolant Systems
undefined
High-Pressure Diaphragm Pumps and Integrated Coolant Systems
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 0,5 NPT(F)
  • 0,5 NPT(F)
  • 0,5 NPT(F)
  • Max. temperature:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • Motor type:
  • TEFC
  • TEFC
  • TEFC
  • Motor hp:
  • 2
  • 2
  • 1
  • Outlet connection:
  • 0,375 NPT(F)
  • 0,375 NPT(F)
  • 0,375 NPT(F)
  • Wetted materials:
  • Brass, Buna-N
  • 316 Stainless Steel, Buna-N
  • 316 Stainless Steel, Buna-N
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together