ContactUsLinkComponent
Masterflex® Doppler Ultrasonic Flowmeters, Avantor®
Masterflex® Doppler Ultrasonic Flowmeters, Avantor®
Certificates
undefined
Masterflex® Doppler Ultrasonic Flowmeters, Avantor®
Specifications
  • Model:
  • 32615-64
  • 32615-66
  • 32615-62
  • 32615-68
  • 32615-60
  • Accuracy:
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • Display type:
  • 2-line x 8-character LCD. Top line (rate): 0,70"; bottom line (totalizer): 0,35"
  • 2-line x 8-character LCD. Top line (rate): 0,70"; bottom line (totalizer): 0,35"
  • 2-line x 8-character LCD. Top line (rate): 0,70"; bottom line (totalizer): 0,35"
  • Min. operating temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Max. operating temperature:
  • 85 °C (185 °F)
  • 99 °C (210 °F)
  • 85 °C (185 °F)
  • 99 °C (210 °F)
  • 85 °C (185 °F)
  • Power:
  • 115 VAC, 50/60 Hz
  • 115 VAC, 50/60 Hz
  • 115 VAC, 50/60 Hz
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year