ContactUsLinkComponent
GF Signet® Paddle-Wheel Flow Sensors
GF Signet® Paddle-Wheel Flow Sensors
Certificates
undefined
GF Signet® Paddle-Wheel Flow Sensors
Specifications
  • Input:
  • 1 to 1000 Hz Frequency
  • 1 to 1000 Hz Frequency
  • 1 to 1000 Hz Frequency
  • 1 to 1000 Hz Frequency
  • Min. operating temperature:
  • –18 °C (0 °F)
  • –18 °C (0 °F)
  • –18 °C (0 °F)
  • –18 °C (0 °F)
  • Max. operating temperature:
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • Max. pressure:
  • 180 psi
  • 180 psi
  • 200 psi
  • 180 psi
  • O-Ring material:
  • FPM
  • FPM
  • FPM
  • FPM
  • Wetted materials:
  • Polypropylene body, black PVDF rotor, Titanium pin, FPM O-rings
  • Polypropylene body, black PVDF rotor, Titanium pin, FPM O-rings
  • Natural PVDF body, rotor and pin, FPM O-rings
  • Polypropylene body, black PVDF rotor, Titanium pin, FPM O-rings
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years