ContactUsLinkComponent
John Guest Polypropylene Push-To-Connect Elbow Adapter Fittings
John Guest Polypropylene Push-To-Connect Elbow Adapter Fittings
Certificates
undefined
John Guest Polypropylene Push-To-Connect Elbow Adapter Fittings
Specifications
  • Collet material:
  • Stainless Steel
  • Stainless Steel
  • Max. operating pressure:
  • 150 psi at 70 °F, 60 psi at 140 °F (for water)
  • 150 psi at 70 °F, 60 psi at 140 °F (for water)
  • Max. temperature:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • Min. temperature:
  • 1 °C (33 °F)
  • 1 °C (33 °F)
  • O-Ring material:
  • EPDM
  • EPDM
  • Sterilisation:
  • Autoclave only
  • Autoclave only
  • Warranty:
  • 20 years
  • 20 years