ContactUsLinkComponent
Masterflex® Adapter Fittings, Male Luer to Threaded, Straight, Avantor®
undefined
Masterflex® Adapter Fittings, Male Luer to Threaded, Straight, Avantor®
  • Model:
  • 45504-84
  • 45518-86
  • 45518-88
  • 45518-82
  • 45518-84
  • Connection type (port 1):
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Connection type (port 2):
  • 10-33
  • ⅛-27 NPT thread
  • ¼-18 NPT thread
  • ¼-28 UNF
  • 10-32 special tapered thread
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • 3 Months
Frequently Bought Together