ContactUsLinkComponent
GoatThroat Spillproof Barrel Pumps, Polypropylene
GoatThroat Spillproof Barrel Pumps, Polypropylene
Certificates
undefined
GoatThroat Spillproof Barrel Pumps, Polypropylene
Specifications
  • Max. temperature:
  • 120 °C
  • 120 °C
  • 120 °C
  • 120 °C
  • Max. viscosity:
  • 1400 SSU
  • 1400 SSU
  • 1400 SSU
  • 1400 SSU
  • Tube length, inlet:
  • 40"
  • 40"
  • 40"
  • 40"
  • Wetted materials:
  • Body: Polypropylene; Seals: Viton
  • Body: Polypropylene; Seals: Santoprene
  • Body: Polypropylene; Seals: EPDM
  • Body: Polypropylene; Seals: Nitrile
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year