ContactUsLinkComponent
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Certificates
undefined
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Specifications
  • Inlet connection:
  • None
  • Tube ext.Ø, outlet:
  • 1,4 cm (0,5625")
  • 1,4 cm (0,5625")
  • 1,4 cm (0,5625")
  • Tube length, inlet:
  • 45,7 cm (18")
  • Tube length, outlet:
  • 121,9 cm (48")
  • 121,9 cm (48")
  • 152,4 cm (60")
  • Wetted materials:
  • PVC barrel and hose, LDPE tube
  • PVC barrel and hose, LDPE tube
  • PVC barrel, shaft, hose