Axypet™ Pro starter kit, Axygen®
Katalognr. AXYGAP-STR-KIT-P
Supplier: Corning
Specifications
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Warsaw
- Pk:1
Specifications
About this item
The Axypet™ Pro starter kit is designed for laboratories which are about to replace their old set of pipettors, and for customers to get a new set of pipettors which cover the most commonly used volumes.
- Three year manufacturer warranty
Certifications: Calibration certificate under ISO8655.
Delivery: Includes four single-channel pipettors: 0.5 to 10 µl (AP-10-P), 2 to 20 µl (AP-20-P), 20 to 200 µl (AP-200-P), 100 to 1000 µl (AP-1000-P); universal linear stand for four single-channel pipettors (AP-USD4-P); Axygen multi rack pipette tips: 10 µl (MR-10XT-R), 200 µl (MR-200-R), 1000 µl (MR-1000XT-R).